BINDER™ Series CB - CO2 Incubator, with Hot Air Sterilization and Heat Sterilizable CO2 Sensor
The powerful and versatile CB CO2 incubator is suitable for cell-based assays in drug and medical research. It is even suitable for complex cultivation experiments or individual environments with hypoxic conditions thanks to its comprehensive assortment of options and accessories.
11530.68 GBP valid until 2025-12-31
Use promo code "25039" to get your promotional price.
Use promo code "25039" to get your promotional price.
Additional Details:
Weight:
121.00000kg
Specification
Capacity (English) | 9.4 cu. ft. |
---|---|
Capacity (English) Shelf | 22 lb. |
Capacity (Metric) Shelf | 10 kg |
Description | CB260-230 V, Standard, Series CB - CO2 incubators with hot air sterilization and heat sterilizable CO2 sensor, 267 L, 230 V 1 ∼ 50/60 Hz |
Capacity (Metric) | 267 L |
Chamber Material | Stainless Steel |
CO2 Concentration Range | 0 to 20% |
CO2 Sensor Technology | IR Sensor |
Convection Technology | Fanless interior |
Convection Type | Natural |
Holds | None |
Lighting | No Lighting |
Material | Stainless Steel |
No. of Places | 1 |
No. of Shelves | 3 |
O2 Concentration Range | 1 to 21% |
Oxygen Control | None |
Plug Type | Nema 5-20P |
Relative Humidity | 90 to 95% |
Temperature Range (Metric) | Ambient +4° to 60°C |
Type | CO2 Incubator |
Display | TFT touch |
Model | CB260-230 V |
O2 Sensor Technology | IR sensor |
Sensor Types | IR sensor |
Dimensions (D x W x H) Interior | 620 x 575 x 750 mm |
Dimensions (L x W x H) | 745 x 885 x 1020 mm |
Weight (Metric) | 125 kg |
Voltage | 230 V |
Wattage | 1.5 kW |
Amperage | 6.5 A |
Frequency | 50/60 Hz |