Use promo code "24135" to get your promotional price.
Thermo Scientific™ SureSTART™ 9 mm Screw Caps, Level 3 High Performance Applications
Use Level 3 Thermo Scientific™ SureSTART™ 9 mm Screw Caps with screw vials that have a 9 mm opening for ultimate performance and sensitivity from your methods, while maintaining the highest levels of sample security and integrity.
389.50 GBP valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Use promo code "24135" to get your promotional price.
Additional Details:
Weight:
0.31350kg
Specification
Color | Black |
---|---|
Material | Polypropylene |
Pre-slit | No |
Septum | White Silicone/Red PTFE |
Level | Level 3 |
Style | Bonded |
Hardness | 54 Shore |
Product Line | SureSTART Level 3 |
Quantity | 1000/Pk. |
Thickness (Metric) | 1 mm |
Certifications/Compliance | Specification Certified |
Type | Screw Cap |
Closure Size | 9 mm |
For Use With (Equipment) | 9 mm Screw Vials |
-
Incredible Savings on Peptide and Protein Mass Spectrometry Testing Products
Explore our new Peptide and Protein Mass Spectrometry Testing Products Promotion
Buy Now