Use promo code "24362" to get your promotional price.
Thermo Scientific™ SureSTART™ 9 mm Screw Caps, Level 3 High Performance Applications
Use Level 3 Thermo Scientific™ SureSTART™ 9 mm Screw Caps with screw vials that have a 9 mm opening for ultimate performance and sensitivity from your methods, while maintaining the highest levels of sample security and integrity.
491.70 GBP valid until 2025-12-31
Use promo code "24362" to get your promotional price.
Use promo code "24362" to get your promotional price.
Additional Details:
Weight:
0.28500kg
Specification
Color | Gray |
---|---|
Material | Polypropylene |
Pre-slit | Yes |
Septum | White Silicone/Red PTFE |
Level | Level 3 |
Style | Pre-slit, Bonded |
Hardness | 54 Shore |
Product Line | SureSTART Level 3 |
Quantity | 1000/Pk. |
Thickness (Metric) | 1 mm |
Certifications/Compliance | Specification Certified |
Type | Screw Cap |
Closure Size | 9 mm |
For Use With (Equipment) | 9 mm Screw Vials |