Memmert™ Entry Port 23 mm for UF160plus, Custom Position TBC Left Side
Optional entry port for Memmert Ovens.
Additional Details:
Additional Details: Weight: 0.01200kg
Product Code. 17509085
Quantity Price
1 £ 166.0 / Each
Estimated Shipment
08-08-2025
Add to basket
Description and Specification
Specification
For Use With (Equipment) UF160plus Oven
Type Entry Port
Diameter (Metric) 23 mm
  • Entry port with 23 mm diameter
  • Flap closure
  • Custom position TBC in left side