Memmert™ Potential-Free Contact (24 V / 2 A) with Socket to NAMUR NE 28 for Signal Generation for UF160plus
Potential-free contact for signal generation, controlled by program segment, for free-selectable functions to be activated.
Additional Details:
Weight:
0.01200kg
Specification
For Use With (Equipment) | UF160plus Oven |
---|---|
Type | Potential-free Contact |
Includes | Socket to NAMUR NE 28 |
Voltage | 24 V |
Amperage | 2 A |