Memmert™ Plug-In Tube Extension for UF160plus, Straight
Plug-in tube extension designed for exhaust air ducting and connection by hose if necessary.
Additional Details:
Additional Details: Weight: 0.01200kg
Product Code. 17519055
Quantity Price
1 £ 110.0 / Each
Estimated Shipment
08-08-2025
Add to basket
Description and Specification
Specification
For Use With (Equipment) UF160plus Oven
Type Plug-in Tube Extension
Diameter (Metric) O.D. 60.3 mm, I.D. 57 mm
  • Accessory
  • Plug-in tube extension (outer diameter 60.3 mm, inner 57 mm)
  • Straight