Memmert™ Entry Port 38 mm for UF160plus
Optional entry port for Memmert Ovens.
Additional Details:
Additional Details: Weight: 0.01200kg
Product Code. 17539085
Quantity Price
1 £ 187.0 / Each
Estimated Shipment
08-08-2025
Add to basket
Description and Specification
Specification
For Use With (Equipment) UF160plus Oven
Type Entry Port
Diameter (Metric) 38 mm
  • Entry port with 38 mm clear diameter
  • Flap and stopper closure
  • Custom position TBC in the back