Memmert™ Interior IP68 Socket for UF160plus
Interior IP68 socket that can be turned on and off with a switch.
Additional Details:
Additional Details: Weight: 0.01200kg
Product Code. 17559095
Quantity Price
1 £ 394.0 / Each
Estimated Shipment
08-08-2025
Add to basket
Description and Specification
Specification
For Use With (Equipment) UF160plus Oven
Type Interior Socket
Voltage 230 V
Amperage 2 A
  • Can only be ordered with limited temperature range - max. +60°C)
  • Electrical Requirements: 230 V, 2 A, 500 W
  • Can be switched off with the On/Off switch
  • Cannot be switched individually
  • Moisture tight IP68
  • Requires option A8 against surcharge