Memmert™ Entry Port 23 mm for UF160plus, Standard Position Left Side Center/Center
Optional entry port for Memmert Ovens.
Additional Details:
Additional Details: Weight: 0.01200kg
Product Code. 17569075
Quantity Price
1 £ 115.0 / Each
Estimated Shipment
08-08-2025
Add to basket
Description and Specification
Specification
For Use With (Equipment) UF160plus Oven
Type Entry Port
Diameter (Metric) 23 mm
  • Entry port with 23 mm diameter
  • Flap and stopper closure
  • Standard position left side center/center