Memmert™ Software AtmoCONTROL for UF160plus
AtmoCONTROL Software meets the requirements for the use of electronically stored data sets and electronic signatures.
Additional Details:
Weight:
0.01200kg
Specification
Certifications/Compliance | Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA) |
---|---|
For Use With (Equipment) | UF160plus Oven |
Type | Software |