Memmert™ Fresh-Air Filter for UF160plus
Fresh-air filter designed to be mounted on the bottom of the unit. Castor frame or subframe not included.
Additional Details:
Additional Details: Weight: 0.01200kg
Product Code. 17569095
Quantity Price
1 £ 265.0 / Each
Estimated Shipment
08-08-2025
Add to basket
Description and Specification
Specification
For Use With (Equipment) UF160plus Oven
Type Fresh Air Filter
  • Requires castor frame or subframe
  • Filtration efficiency 80%
  • Mounted at the bottom of the appliance