Memmert™ Stainless Steel Bottom Drip Tray for UF160plus, 15 mm
Non-perforated bottom drip trays constructed of stainless steel.
Additional Details:
Additional Details: Weight: 0.01200kg
Product Code. 17589045
Quantity Price
1 £ 172.0 / Each
Estimated Shipment
08-08-2025
Add to basket
Description and Specification
Specification
For Use With (Equipment) UF160plus Oven
Type Bottom Drip Tray
Material Stainless Steel
Height (Metric) Rim: 15 mm
  • Accessory
  • May affect temperature distribution
  • Constructed of stainless steel
  • Rim height: 15 mm