Thermo Scientific™ Multidrop™ Combi Reagent Dispenser
Meet your lab's specific needs with outstanding flexibility and performance with the easy-to-use Thermo ScientificTM MultidropTM Combi Reagent Dispenser.
Overview
15916.20 GBP valid until 2025-06-30
Use promo code "25076" to get your promotional price.
Additional Details:
Additional Details: Weight: 14.25000kg
Product Code. 17801692
Quantity Price
1 £ 22375.0 / Each
In stock, ready to ship: 5
Add to basket
  • Description and Specification
  • Alternative Products
  • Special Offers
Description and Specification
Specification
Accuracy Small tube dispensing cassette: 2μL, ±10%; 10μL, ±5% Standard tube dispensing cassette: 20μL, ±2%; 100μL, ±1%
Product Type Reagent Dispenser
Description Multidrop™ Combi+ Reagent Dispenser
Channels 8
Volume (Metric) 0.5 to 2500 μL
For Use With (Application) The instrument is intended for automated dispensing of up to eight different reagents simultaneously into 6, 12, 24, 48, 96, or 384-well plate format. It can be used in several applications, for example, in drug discovery/high-throughput screening, genomic, proteomic and cell-based assays, and ELISA. It is capable of dispensing into 2D tubes in SLAS format that are shorter than 50mm.  
For Use With (Equipment) Can be used with automated systems (e.g. Thermo Scientific™ Orbitor™ Microplate Mover) and a variety of microplates and strips in 96, 384 or 1536-well plate format
Includes One standard tube dispensing cassette (cat. no. 24072670)
Interface On-board software and Serial RS-232 or USB for PC control
Plate Type 6-1536 - 6, 12, 24, 48, 96, and 384 well plates
Precision
  • Small tube dispensing cassette: 2 μL: CV ≤ 5%; 10 μL: CV ≤ 3% 
  • Standard tube dispensing cassette:  20 μL: CV ≤ 1.5%; 100 μL: ≤ CV 1% 
Speed
  • 96-well plate: 10μL in 5 sec.; 20μL in 5sec.; 100μL in 10 sec 
  • 384-well plate: 1μL in 6 sec.; 5μL in 9 sec.; 10μL in 13 sec.; 20μL in 21 sec. 
  • 1536-well plate: 1μL in 15 sec.; 5μL in 28 sec. 
Volume (English)
  • Small tube dispensing cassette:  0.5-50μL, in 0.5μL increment 
  • Standard tube dispensing cassette:  5-2500μL, in 5 μL increment 
Volume (Metric) Dead <1mL volume range 0.5-50μL; <7mL volume range >50μL (it is possible to blackflush the reagent into original reservoir to avoid loss of expensive reagent)
Warranty 1 year
Dimensions (L x W x H) 14 x 8.6 x 12.9 in.; 35.5 x 22 x 33 cm
Weight (English) 20.1 lb 
Weight (Metric) 9.1 kg 
 The Multidrop Combi Reagent Dispenser handles a wide selection of plates and volume range and provides fast dispensing and high-throughput operation.
  • Provides precise dispensing over a 0.5 to 2500μL range, ensuring reproducible assay data
  • Accommodates microplates from 6 to 1536 wells and plate heights of 5 to 50mm
  • Visual icon-based graphic display makes it easy to use and program, even without training
  • Minimal dead volume and back-flushing reduces reagent costs
  • Easy to use Thermo Scientific FILLit Software provides increased flexibility and functionality
  • Full robotic compatibility gives increased throughput
  • Uses eight-channel detachable and autoclavable dispensing cassettes that are standard across the Multidrop range
Multidrop Combi SMART:
  • Adds an advanced built-in tracing system to the Multidrop Combi
  • Provides improved realiability and cassette lifetime traceability, greatly enhancing user efficiency and reporting capability
Recommended for:
  • Assay development
  • Primary and secondary screening
  • Compound storage
  • Genomics and proteomics research
  • PCR set-up
  • Sequencing set-up
  • Cell based assays
  • Bead based assays
  • ELISA assays