Brannan™ LO-tox™ Lab Thermometer with 3-points UKAS Calibration
LO-tox™ lab thermometer complete with calibration certificate.
Additional Details:
Weight:
0.03900kg
Specification
Calibration Points | 0°C, 25°C, 45°C |
---|---|
Certifications/Compliance | ISO 17025 UKAS |
Immersion Type | Total Immersion |
Length (Metric) | 40.5 cm |
Material | Glass |
Thermometer Fill | Blue Spirit |
Max. Temperature (English) | 124°F |
Max. Temperature (Metric) | +51°C |
Min. Temperature (English) | 30°F |
Min. Temperature (Metric) | -1°C |
Temperature Range (English) | 30°F to 124°F |
Temperature Range (Metric) | -1°C to 51°C |
Accuracy | ± 0.5°C at 35°C |
Subdivision | 0.2°C |
Width (Metric) | 1.2 cm |