BINDER™ Series FP Avantgarde.Line - Drying and Heating Chambers with Forced Convection and Program Functions
A BINDER material test chamber with mechanical convection of the FP series provides reliably short drying times and particularly fast heating – even for chambers under full loads.
2511.55 GBP valid until 2025-06-30
Use promo code "24075" to get your promotional price.
Use promo code "24075" to get your promotional price.
Additional Details:
Weight:
55.00000kg
Specification
Capacity (Metric) | 116 L |
---|---|
Temperature (Metric) | 300°C |
Type | Drying and Heating Chamber |
Display | LED |
Model | FP115-230V |
Dimensions (D x W x H) Interior | 383 x 550 x 550 mm |
Dimensions (L x W x H) | 710 x 636 x 732 mm |
Weight (Metric) | 62 kg |
Voltage | 230 V |
Wattage | 1.6 kW |
Amperage | 10 A |
Frequency | 50/60 Hz |
-
The Ultimate Lab Appliances Promotion
Enjoy discounts up to 78% on top-quality lab equipment and upgrade your laboratory with the best appliances at the lowest prices.
Buy Now