Brannan™ LO-tox™ Lab Thermometer with 3-points Traceable Calibration
LO-tox™ lab thermometer complete with calibration certificate.
Additional Details:
Additional Details: Weight: 0.03900kg
Product Code. 17862988
Quantity Price
1 £ 139.36 / Each
Estimated Shipment
04-07-2025
Specification
Calibration Points 0°C, 50°C, 90°C
Certifications/Compliance UK Traceable to National Standards ISO 9001
Immersion Type Partial Immersion
Length (Metric) 40.5 cm
Material Glass
Thermometer Fill Blue Spirit
Max. Temperature (English) 214°F
Max. Temperature (Metric) +101°C
Min. Temperature (English) 30°F
Min. Temperature (Metric) -1°C
Temperature Range (English) 30°F to 214°F
Temperature Range (Metric) -1°C to 101°C
Accuracy ± 1.0°C at 90°C
Subdivision 0.2°C
Immersion Length 76 mm
Width (Metric) 1.2 cm
  • LO-tox™ is an organic filling specially chosen to achieve a wide temperature span and provide the lowest practical levels of hazard
  • Safe non-mercury filling
  • Supplied with calibration certificate
  • Non-roll end cap
  • Supplied as standard in protective individual, resealable telescopic tube
  • Made in UK
Alternative Products