76.42 GBP valid until 2025-08-24
Use promo code "24132" to get your promotional price.
Use promo code "24132" to get your promotional price.
Certifications/Compliance | OSHA, 29 CFR1910.1030 |
---|---|
Closure Type | Elastic |
Color | Clear with Red Print |
Dimensions (L x W) | 24 x 12 in. (61 x 30 cm) |
Length (Metric) | 61 cm |
Width (Metric) | 30 cm |
Autoclavable | Autoclavable |
Includes | Printed in English and Spanish |
Width (English) | 12 in. |
Length (English) | 24 in. |
Thickness | 1.50 mil |
Material | Polyethylene |
Product Type | Autoclavable Bags |
Temperature (Metric) | 127°C |