Use promo code "24112" to get your promotional price.
Thermo Scientific™ PCR Plate, 96-well, segmented, semi-skirted, Sustain™ Series
The Thermo Scientific PCR Plate, 96-well, segmented, semi-skirted, Sustain Series, provides superior rigidity and space for barcoding. The patented segmented design of this plate allows it to be cut into 24- and 48-well sections.
75.33 GBP valid until 2025-06-21
Use promo code "24112" to get your promotional price.
Use promo code "24112" to get your promotional price.
Additional Details:
Weight:
0.68000kg
Specification
Barcode | Custom Barcoding Available |
---|---|
Certifications/Compliance | Certified DNase-, RNase-, Human DNA-, and PCR Inhibitor-free |
Format | SBS Standard |
Autoclavable | Autoclavable |
Color | Natural |
Skirt Style | Semi |
Material | Polypropylene |
No. of Wells | 96 |
Green Features | Less waste, renewable material |
Quantity | 25 plates |
-
Freedom of Expression
Let your genes speak volumes with qPCR master mixes made for high-stakes science.
Buy Now