Use promo code "24112" to get your promotional price.
Thermo Scientific™ PCR Plate, 96-well, low profile, skirted, Sustain™ Series
The Thermo Scientific PCR Plate, 96-well, low profile, skirted, Sustain Series, has a low-profile, thin well wall design and is compatible with PCR and qPCR applications.
78.00 GBP valid until 2025-06-21
Use promo code "24112" to get your promotional price.
Use promo code "24112" to get your promotional price.
Additional Details:
Weight:
0.73000kg
Specification
Barcode | Custom Barcoding Available |
---|---|
Certifications/Compliance | Certified DNase-, RNase-, Human DNA-, and PCR Inhibitor-free |
Format | SBS Standard |
Autoclavable | Autoclavable |
Color | Natural |
Skirt Style | Skirted |
Material | Polypropylene |
No. of Wells | 96 |
Green Features | Less waste, renewable material |
Quantity | 25 plates |