Proteintech ATG2A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Additional Details:
Additional Details: Weight: 0.00010kg
Product Code. 16862245
Quantity Price
1 £ 135.0 / 20µL
In stock, ready to ship: 1
Add to basket
Description and Specification
Specification
Antigen ATG2A
Applications Western Blot,Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ATG2A
Gene Accession No. Q2TAZ0
Gene Alias ATG2A, KIAA0404
Gene Symbols ATG2A
Host Species Rabbit
Immunogen ATG2A Fusion Protein Ag19709
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23130
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Required for both autophagosome formation and regulation of lipid droplet morphology and dispersion.