Use promo code "25339" to get your promotional price.
Thermo Scientific Acros Benzaldehyde, 98+%, pure, Thermo Scientific Chemicals
Use promo code "25339" to get your promotional price.
Quantity | 2.5 L |
---|
Chemical Identifiers
CAS | 100-52-7 |
---|---|
Molecular Formula | C7H6O |
Molecular Weight (g/mol) | 106.12 |
InChI Key | HUMNYLRZRPPJDN-UHFFFAOYSA-N |
IUPAC Name | benzaldehyde |
SMILES | O=CC1=CC=CC=C1 |
Specification
Chemical Name or Material | Benzaldehyde |
---|---|
CAS | 100-52-7 |
Melting Point | -26.0°C |
Density | 1.0430 g/mL |
Boiling Point | 179.0°C |
Flash Point | 64°C |
Assay Percent Range | 98+% |
Packaging | Glass bottle |
Molecular Formula | C7H6O |
Linear Formula | C6H5CHO |
Refractive Index | 1.5440 to 1.5460 |
Quantity | 2.5 L |
Beilstein | 07, 174 |
Fieser | 01,46; 06,26; 13,108 |
Merck Index | 15, 1060 |
Specific Gravity | 1.043 |
Solubility Information | Solubility aq. soln.: slightly soluble. Other solubilities: miscible with alcohol, ether and most other common, organic solvents |
InChI Key | HUMNYLRZRPPJDN-UHFFFAOYSA-N |
SMILES | O=CC1=CC=CC=C1 |
IUPAC Name | benzaldehyde |
Molecular Weight (g/mol) | 106.12 |
Formula Weight | 106.12 |
Grade | Pure |
Technical documents
[Benzaldehyde]
[Warning]
[{code=ACUTE TOX. 4, sequence=1, description=Acute toxicity Category 4}, {code=EYE DAM. 2, sequence=2, description=Serious eye damage/eye irritation Category 2}, {code=STOT SE 3, sequence=3, description=Specific target organ toxicity Category 3}, {code=AQUA CHRONIC 3, sequence=4, description=LONG-TERM AQUATIC HAZARD Chronic 3}, {code=SKIN IRRIT. 2, sequence=5, description=Skin corrosion/irritation Category 2}]
[{code=H319, sequence=1, description=Causes serious eye irritation.}, {code=H335, sequence=2, description=May cause respiratory irritation.}, {code=H332, sequence=3, description=Harmful if inhaled.}, {code=H302, sequence=4, description=Harmful if swallowed.}, {code=H315, sequence=5, description=Causes skin irritation.}, {code=H412, sequence=6, description=Harmful to aquatic life with long lasting effects.}]
[{code=P280, sequence=50, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P301+P310, sequence=51, description=IF SWALLOWED: Immediately call a POISON CENTER/doctor /}, {code=P302+P352, sequence=53, description=IF ON SKIN: Wash with plenty of water/.}, {code=P304+P340, sequence=54, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P305+P351+P338, sequence=55, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P312, sequence=56, description=Call a POISON CENTER/doctor/if you feel unwell.}]
[{code=GHS07, sequence=1, description=EXCLAMATION MARK}]
Product Identifier
- Benzaldehyde
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
- Skin corrosion/irritation Category 2
Hazard Statement
- H319- Causes serious eye irritation.
- H335- May cause respiratory irritation.
- H332- Harmful if inhaled.
- H302- Harmful if swallowed.
- H315- Causes skin irritation.
- H412- Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310- IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352- IF ON SKIN: Wash with plenty of water/.
- P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312- Call a POISON CENTER/doctor/if you feel unwell.