Code:
NEW
Axygen™ Black Hanging Format Automation Pipet Tips for Hamilton Microlab™ Star Line and Nimbus™, 5 Pack
Compatible with Hamilton Microlab™ Star Line and Nimbus™ automated liquid handling workstations
Additional Details:
Weight:
4.28000kg
Specification
Volume (Metric) | 50 μL |
---|---|
Filtered | Filtered |
Sterility | Sterile |
Color | Black |
Autoclavable | Not Autoclavable |
Clean Claims | DNase, RNase and Pyrogen free |
For Use With (Application) | Available in four sizes and specially designed for use with Hamilton™ liquid handling platforms, they enable precision pipetting across a wide range of applications |
Format | Hanging Tip Rack |
Tip Style | Automation |
Packaging | 5 Racks/Pk, 12 Pks/Cs. |
For Use With (Equipment) | Hamilton MICROLAB Nimbus |
Material | Polypropylene |