Hirschmann™ Class AS Glass Burettes, Lateral Stopcock with PTFE-Spindle
Excellent for automatically dispensing measured amounts of chemical solutions in the laboratory. Hirschmann Instruments™ Class AS Glass Burets, Lateral Stopcock with PTFE-Spindle are manufactured from durable DURAN™ glass tubing.
Additional Details:
Weight:
0.25000kg
Specification
Capacity (Metric) | 50 mL |
---|---|
Material | Glass |
Graduations | 0.1 mL |
Includes | Schellbach stripes |
Scale | Blue |
Stopcock Material | PTFE |
Tolerance | 0.05 |
Type | Buret |
Class | Class AS |
Certifications/Compliance | DIN EN ISO 385 |