Agilent BioTek Blue Test Dye, 10X Concentrate
For use with ELx405 and ELx50 Microplate Washers
Additional Details:
CAS Number:
9005-64-5
Weight:
0.12000kg
Specification
Product Type | Blue Test Dye, 10X Concentrate |
---|---|
For Use With (Equipment) | Agilent BioTek Microplate Washers and Readers |
Quantity | 125 mL |