Reagecon 1LT BUFFER PH4.00 (RED) 0.01 25°C 1L
Overview
1LT BUFFER PH4.00 (RED) 0.01 25°C 1L
Code: NEW
Additional Details:
Additional Details: Weight: 0.99000kg
Product Code. 11434579
Quantity Price
1 £ 47.0 / 1 L
In stock, ready to ship: 118
In stock, ready to ship: 24
Add to basket
  • Alternative Products