Proteintech C4BPA Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Additional Details:
Weight:
0.00010kg
Specification
Antigen | C4BPA |
---|---|
Applications | Immunohistochemistry (Paraffin),Western Blot |
Classification | Polyclonal |
Concentration | 0.27 mg/mL |
Conjugate | Unconjugated |
Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
Gene | C4BPA |
Gene Accession No. | P04003 |
Gene Alias | C4BP, C4BPA, Proline rich protein, PRP |
Gene Symbols | C4BPA |
Host Species | Rabbit |
Immunogen | C4BPA Fusion Protein Ag2387 |
Purification Method | Antigen Affinity Chromatography |
Quantity | 150 μL |
Regulatory Status | RUO |
Primary or Secondary | Primary |
Gene ID (Entrez) | 722 |
Target Species | Human |
Content And Storage | -20°C |
Product Type | Antibody |
Form | Liquid |
Isotype | IgG |