BINDER™ Calibration Certificate, Temperature
Calibration Certificate for temperature measurement.
Additional Details:
Weight:
1.00000kg
Specification
For Use With (Equipment) | BINDER Units M 56, M 115, M 260, M 720 |
---|---|
Description | Expanded temperature, for extending the measurement in center of chamber to include another test temperature |
Calibration Points | 1 Expanded Temperature Measurement Point (Center Chamber) |
Duration | 1 Day |
Product Type | Calibration Certificate (Expanded) |
Service Offering | 1 Year Extension, No Inspection |