Fisherbrand™ Catalyst Norprene Food Formulation Stopped Tubing Assembly
Preconfigured ready-to-use links and assemblies.
Additional Details:
Additional Details: Weight: 0.45000kg
Product Code. 15501345
 N/A
N/A
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description and Specification
Specification
Description Stopped tubing assembly
Diameter (Metric) Inner 9.5 mm
Thickness (Metric) 2.4 mm
  • Available in Platinum-Cured Silicone, TPEs, and PVC
  • Ensures optimal amount of tubing is in pump head