Fisherbrand™ Catalyst Norprene Food Formulation Stopped Tubing Assembly
Preconfigured ready-to-use links and assemblies.
Additional Details:
Weight:
0.45000kg
Specification
Description | Stopped tubing assembly |
---|---|
Diameter (Metric) Inner | 4.8 mm |
Thickness (Metric) | 1.6 mm |