Thermo Scientific™ Decappers 500/550, 12-Channel
Get automated decapping and capping of Thermo Scientific;trade; screw top tubes in ANSI racks with Quick Switch Technology for medium to high throughput laboratories.
Additional Details:
Weight:
0.50000kg
Specification
| Compatible Storage Tubes | Nunc Cryobank Tubes (0.5 and 1 mL) |
|---|---|
| Decapping Format | 96-Channel |
| Description | Adapter kit, 96 Nunc |
| Type | Accessory |
| For Use With (Equipment) | Decapper 500/550 |