Velp Scientifica™ FOC 200E Connect Cooled Incubator
Refrigerated thermostat of 200 L that guarantees excellent connectivity and high performance in temperature control with the Auto-Tuning System.
Overview
1808.08 GBP valid until 2025-06-30
Use promo code "24075" to get your promotional price.
Additional Details:
Additional Details: Weight: 43.00000kg
Product Code. 16885233
Quantity Price
1 £ 1950.0 / Each
Estimated Shipment
14-07-2025
Add to basket
  • Description and Specification
  • Alternative Products
  • Special Offers
Description and Specification
Specification
Capacity (English) 44.6 gal.
Capacity (English) Shelf 22 lb.
Capacity (Metric) Shelf 10 kg
Certifications/Compliance CE Declaration of Conformity
Color White
Autoclavable Not Autoclavable
Capacity (Metric) 169 L
Chamber Material Plastic
Lighting No Lighting
No. of Shelves 3
Oxygen Control None
Plug Type EU, UK Optional
Temperature Range (Metric) 3°C to 50°C
Display Digital, 3 Digit
Frame Material Metal
No. of Doors 1
Sterilization Indicator No
Temperature Accuracy ±0.5°C
Temperature Resolution ±0.5°C
Temperature Sensitivity ±0.5°C
Warranty 25 Months
Dimensions (D x W x H) Interior 550 x 540 x 1263 mm
Height (English) 50 in.
Height (English) Interior 3.5 ft.
Height (Metric) 1263 mm
Height (Metric) Interior 1070 mm
Depth (English) 21.7 in.
Depth (English) Interior 13.8 in.
Depth (Metric) 550 mm
Depth (Metric) Interior 350 mm
Weight (English) 110.2 lb.
Weight (Metric) 50 kg
Electrical Requirements 230 V, 50/60 Hz
Voltage 230 V
Frequency 50/60 Hz
Thermo Regulation and Stability:
  • Auto-Tuning thermoregulation system ensures optimum thermal homogeneity and stability (± 0.5°C).
  • Selectable incubation temperature and working range from 3°C to 50°C.
  • Uniform temperature thanks to VELP ventilation technology.
  • Eco-friendly cooling system.
  • Low operating costs.
Reliable and Easy to Control:
  • Intuitive digital interface, with three keys for easy setup of the instrument.
  • Accepts up to 4 VELP Respirometer systems (4 BOD Sensor System 6, or 1 BOD Sensor System 10).
  • Internal sockets to power devices.
  • Constant monitoring according to GLP (Good Laboratory Practice).
  • Adjustable recording time (in case of connection to VELP Ermes and TempSoft™).
  • Optional TempSoft™ incorporates high/low alarm thresholds.
  • Quick and easy calibration.
  • Total volume of 200 L
Monitor with Velp Ermes:
  • Reduce daily manual tasks and repetitive operations with real-time control of the FOC Connect from anywhere.
  • Data Logging and Insights on the instrument are available to enhance productivity and reproducibility.
  • Immediate notifications and alerts will allow users to always be informed about the status of their analysis.
  • Benefit from software updates and immediate ad hoc assistance, without the need for specialized technicians.
Control Through TEMPSoft™ Software:
  • Control up to 10 FOC Connect at once.
  • Set working ramps (different times and temperatures) and the desired setpoint.
  • Monitor internal temperature with a dedicated graph.
  • Export data into multiple formats.
  • Store test data in conformity with GLP (Good Laboratory Practice).
Alternative Products
Special Offers