200.14 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Use promo code "25339" to get your promotional price.
Capacity (Metric) | 100 mL |
---|---|
Color | Clear-Red |
Material | Glass, Rubber |
Quantity | 1 Ea. |
Type | Laboratory Sprayer |
For Use With (Application) | TLC |
Includes | Rubber bulb/tubing |