Invitrogen™ Anza™ 10-Pack Starter Kit
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Overview
318.25 GBP valid until 2025-12-31
Use promo code "24368" to get your promotional price.
Code: NEW
Additional Details:
Additional Details: Weight: 0.50000kg
Disclaimers: For Research Use Only. Not for use in diagnostic procedures.
Includes: Anza™ 11 EcoRI, Anza™ 5 BamHI, Anza™ 1 NotI, Anza™ 16 HindIII, Anza™ 12 XbaI, Anza™ 3 BcuI, Anza™ 8 XhoI, Anza™ 10 DpnI, Anza™ 19 BglII, Anza™ 26 Eco32I, Anza™ T4 DNA Ligase Master Mix, Anz
Product Code. 15441933
Quantity Price
1 £ 489.0 / 1 set
Estimated Shipment
18-02-2026
Add to basket
  • Description and Specification
  • Alternative Products
Description and Specification
Specification
Content And Storage 20 μL each of Anza 11 EcoRI, Anza 5 BamHI, Anza 1 NotI, Anza 16 HindIII, Anza 12 XbaI
20 μL each of Anza 3 BcuI, Anza 8 XhoI, Anza 10 DpnI, Anza 19 BglII, Anza 26 Eco32I
100 μL Anza T4 DNA Ligase Master Mix
20 μL Anza Alkaline Phosphatase
500 μL Anza Buffer (10X)
500 μL Anza Red Buffer (10X)
Store at -5 to -30°C.
Shipping Condition Approved for shipment on Wet or Dry Ice
Methylation Sensitivity Methylation Sensitivities Varies by Enzyme
Enzyme HindIII, NotI, Alkaline Phosphatase, XhoI, BcuI, XbaI, DpnI, T4 DNA Ligase, Eco32I, BamHI, BglII, EcoRI
Compatible Buffer Anza™ 10X Red Buffer, Anza™ 10X Buffer
Type IIS RE No
Quantity 1 kit
Product Type Starter Kit
Research Category Traditional Cloning
Product Line Anza
The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.
  • One buffer for all restriction enzymes
  • One digestion protocol for all DNA substrates
  • Complete digestion in 15 min
  • Includes 10 common restriction enzymes
  • Complete with Anza T4 DNA Ligase Master Mix and Anza Alkaline Phosphatase
Certifications: CoA
Alternative Products