Implen™ NanoPhotometer™ 21 CFR Part 11 Software Packages
CFR21 Software complies with FDA 21 CFR Part 11 requirements, and is an optional software tool that is ideal for GxP laboratories which require proper electronic record keeping.
Additional Details:
Weight:
0.10000kg
Disclaimers:
For Research Use Only
Specification
| Certifications/Compliance | FDA 21 CFR Part 11 |
|---|---|
| Description | Software Upgrade for NanoPhotometer |
| For Use With (Equipment) | NanoPhotometer™ N120, NP80, N60, and C40 |
| Includes | Software License Key |
| System Requirements | Windows or Mac, Touchscreen Compatible |