Cytiva Ni Sepharose™ 6 Fast Flow
Specification
Buffer | 0.1M HCl, 0.1M NaOH, 8M Urea |
---|---|
Product Type | Ni Sepharose 6 FF chromatography media |
Quantity | 500 mL |
Height (English) | 9.84 in. |
Height (Metric) | 250mm |
Flow Rate | 600 cm/hr. |
Max. Pressure | 1 bar |
pH Range | 3 to 12 |
Particle Size | 90μm |
Support Type | 6% Cross-linked Agarose |
Content And Storage | 4° to 30°C |
dimMatrix | Ni-Sepharose |
[Warning]
[{code=FLAM. LIQ. 3, sequence=1, description=Flammable liquid Category 3}, {code=AQUATIC ACUTE 1, sequence=2, description=ACUTE AQUATIC Acute 1}, {code=AQUA CHRONIC 3, sequence=3, description=LONG-TERM AQUATIC HAZARD Chronic 3}]
[{code=H226, sequence=1, description=Flammable liquid and vapour.}, {code=H400, sequence=2, description=Very toxic to aquatic life.}, {code=H412, sequence=3, description=Harmful to aquatic life with long lasting effects.}]
[{code=P210, sequence=51, description=Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.}, {code=P241, sequence=52, description=Use explosion-proof electrical/ventilating/lighting/ equipment.}, {code=P273, sequence=53, description=Avoid release to the environment.}, {code=P501b, sequence=54, description=Dispose of contents/container in accordance with local/regional/national/international regulations.}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS09, sequence=2, description=ENVIRONMENT}]
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H226- Flammable liquid and vapour.
- H400- Very toxic to aquatic life.
- H412- Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241- Use explosion-proof electrical/ventilating/lighting/ equipment.
- P273- Avoid release to the environment.
- P501b- Dispose of contents/container in accordance with local/regional/national/international regulations.