Miele Professional™ Injector nozzle, Jet A 802
Jet A 802 for the optimum cleaning of the dispenser compartment
Overview
49.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Code: NEW
Additional Details:
Additional Details: Weight: 0.02700kg
Product Code. 15431581
Quantity Price
1 £ 43.8 / Each
Estimated Shipment
30-04-2026
Add to basket
Description and Specification
Specification
Depth (Metric) 15 mm
Height (Metric) 187 mm
Width (Metric) 30 mm
  • For use with lab washers with door powder dispensing
  • For use with injector modules to rinse out powder residue from door dispenser