36.15 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Use promo code "25339" to get your promotional price.
| Backing Material | Polyester |
|---|---|
| Material | Unmodified Silica Gel (SiOH) |
| Particle Size | 5 to 17 μm |
| Pore Size | 60 Å |
| Quantity | 50/Pk. |
| Type | Polyester Sheet |
| Pore Volume | 0.75 mL/g |
| Sorbent | Unmodified Silica Gel (SiOH) |
| Thickness (Metric) | 0.2 mm |
| Dimensions (L x W) | 4 x 8 cm |