Macherey-Nagel™ Polygram SIL G Polyester Sheets
Macherey-Nagel Polygram SIL G Polyester Sheets, Dimensions: 40 x 20 cm, Pore Size: 60 A, Pore Volume: 0.75 mL/g, Quantity: 25 Pack, Thickness: 0.2 mm
Overview
82.55 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Code: 16
Additional Details:
Additional Details: Weight: 0.50000kg
Product Code. 10709182
Quantity Price
1 £ 303.0 / Pack of 25
In stock, ready to ship: 68
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
Description and Specification
Specification
Backing Material Polyester
Material Unmodified Silica Gel (SiOH)
Particle Size 5 to 17 μm
Pore Size 60 Å
Quantity 25/Pk.
Type Polyester Sheet
Pore Volume 0.75 mL/g
Sorbent Unmodified Silica Gel (SiOH)
Thickness (Metric) 0.2 mm
Dimensions (L x W) 20 x 20 cm
Alternative Products
Documentation