Applied Biosystems™ QuantStudio™ Absolute Q Package, including Digital PCR System, desktop PC, Smart Start training and 1 Year extended warranty
The Applied Biosystems™ QuantStudio™ Absolute Q™ Digital PCR System combines the power of digital PCR (dPCR) absolute quantification with the simplicity of a qPCR workflow. With a single hands-on step that takes five minutes to complete, dPCR is now easier than ever.
Additional Details:
Weight:
21.00000kg
Specification
| Block Format | Non-interchangeable |
|---|---|
| Content And Storage | Digital PCR System, Desktop PC, Smart Start Training and 1 Year Extended Warranty |
| For Use With (Equipment) | QuantStudio™ Absolute Q™ MAP16 Plate |
| Product Line | QuantStudio |
| Product Type | Digital PCR System |
| For Use With (Application) | Digital PCR |
| Shipping Condition | Room Temperature |