Proteintech RBM3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Additional Details:
Additional Details: Weight: 0.00010kg
Product Code. 16842974
Quantity Price
1 £ 349.0 / 150µL
In stock, ready to ship: 3
Add to basket
Description and Specification
Specification
Antigen RBM3
Applications Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot
Classification Polyclonal
Concentration 0.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RBM3
Gene Accession No. O89086, P98179, Q925G0
Gene Alias IS1 RNPL, Putative RNA binding protein 3, RBM3, RNA binding motif protein 3, RNPL
Gene Symbols RBM3
Host Species Rabbit
Immunogen RBM3 Fusion Protein Ag5934
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114488, 19652, 5935
Target Species Human,Mouse,Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
This gene is a member of the glycine-rich RNA-binding protein family and encodes a protein with one RNA recognition motif domain. Expression of this gene is induced by cold shock and low oxygen tension. A pseudogene exists on chromosome 1. Multiple alternatively spliced transcript variants that are predicted to encode different isoforms have been characterized although some of these variants fit nonsense-mediated decay criteria.