Memmert™ Reinforced Stainless Steel Shelf
Additional reinforced stainless steel shelf for reinforced chamber option
Additional Details:
Additional Details: Weight: 0.01100kg
Product Code. 17170045
Quantity Price
1 £ 625.0 / Each
Estimated Shipment
05-08-2025
Add to basket
Description and Specification
Specification
For Use With (Equipment) IPP1060eco
Material Stainless Steel
  • With guide bars and fixing screws
  • Requires option K1
  • Consider max. loading of chamber; max. loading 60 kg