6.16 GBP valid until 2025-12-31
Use promo code "24380" to get your promotional price.
Use promo code "24380" to get your promotional price.
Product Type | Hooded Coveralls |
---|---|
Size | Small |
Material | Flashspun HDPE |
Color | White |
Disposable | Single-use |
Waist Style | Elastic at Back |
Ankle Style | Elastic |
Cuff Style | Elastic |
Neck Style | Hooded |
Hood Type | 3-panel |
Closure Type | Self-locking Zipper with Flap |
Seam Type | Stitched |
Certifications/Compliance | Category III, Type 5-B and 6-B, EN 14126, EN 1073-2, EN 1149-5 |
Size (Metric) Chest | 84 to 92 cm |
For Use With (Application) | Pharmaceutical handling, chemical processing, the oil and gas industry, general maintenance and operations, automatic spray painting and many others. |
Quantity | 100/Pk. |
Style | Classic Xpert |