OHAUS™ Support Stand with Rod
OHAUS™ Support Stand with Rod provides secure support for holding glassware at elevated positions.
Overview
175.49 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Additional Details:
Additional Details: Weight: 0.50000kg
Product Code. 15762359
Quantity Price
1 £ 234.0 / Each
Estimated Shipment
25-02-2026
Add to basket
Description and Specification
Specification
Type Support Stand
Material Aluminum
Includes Rod
Description Clamp, Specialty, Stand/Rod, CLS-STRODA
Diameter (Metric) 13 mm Rod
Height (Metric) 25.000 mm
Height (English) 1.00 in.
Length (English) 7.00 in.
Length (Metric) 178 mm
Length (Metric) Arm 578 mm
Width (English) 13.00 in.
Width (Metric) 330.00 mm
  • Corrosion-resistant lab-frames, rods and connectors are available in premium stainless steel, offering high chemical resistance and durability. Aluminum offers reliable strength at a lighter weight.
  • Lab lifts offer convenient adjustable lifting platforms with exceptional strength and durability for holding small lab equipment such as glassware, hot plates and water baths at different heights.
  • Simplify lab set up! Thirteen lab-frame kit options are available with four designed to fit in fume hoods. Build your own support system choosing from a wide selection of rods, bases and connectors.
  • Dependable connectors secure rods and clamps to meet your requirements and five unique holders in a variety of sizes and materials accommodate different rods.