171.00 GBP valid until 2025-12-31
Use promo code "24350" to get your promotional price.
Use promo code "24350" to get your promotional price.
Material (Liner) | PTFE/14B Rubber |
---|---|
Color | Clear |
Diameter (Metric) Outer | 28 mm |
Material | Borosilicate Glass (Low Extractable) |
Volume (Metric) | 20 mL |
For Use With (Application) | Low profile for greater stability and less dead volume above the sample, making it ideal for compound storage |
Thread Style | Screw |
Volume (English) | 5 dr. |
Closure Material | Phenolic |
Autoclavable | Autoclavable |
Closure Type | Screw Cap |
Height (Metric) | 60 mm |
Certifications/Compliance | ASTM Type I, Class A, USP Type I |
Closure Size | 24-400 |
For Use With (Equipment) | M-T File™ and Vial Rack |