Cytiva Whatman™ Cellulose Nitrate Membrane Filters
Manufactured under strictly controlled conditions. Whatman™ Cellulose Nitrate Membrane Filters have a noticeably improved flexibility and are made to tolerate abuse during handling, loading, and autoclaving without sacrificing integrity.
Overview
Code: 69
Additional Details:
Additional Details: Weight: 0.03000kg
Product Code. 10353101
Quantity Price
1 £ 269.0 / Pack of 100
In stock, ready to ship: 52
Add to basket
  • Description and Specification
  • Alternative Products
  • Safety and Handling
Description and Specification
Specification
Format Disc
Material Cellulose Nitrate
Pore Size 5 μm
Quantity 100 Pk.
Thickness (Metric) 105 to 140 μm
Diameter (English) 1.85 in.
Diameter (Metric) 47 mm
Among the strongest of their type available, as measured and compared by burst pressure tests.
  • Narrow pore size distribution for improved surface capture and analysis.
  • Low levels of extractables to ensure sample integrity.
  • High strength and flexibility
  • Increased temperature stability
  • Reduced shrinkage
Alternative Products
Safety and Handling
[X100 MEMBRANFILTER AE98 47MM,]
[Danger]
[{code=FLAM. SOL. 2, sequence=1, description=Flammable solid Category 2}]
[{code=H226, sequence=1, description=Flammable liquid and vapour.}]
[{code=P210, sequence=51, description=Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.}, {code=P241, sequence=52, description=Use explosion-proof electrical/ventilating/lighting/ equipment.}, {code=P260, sequence=53, description=Do not breathe dust/fume/gas/mist/vapours/spray.}, {code=P280, sequence=54, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P501b, sequence=55, description=Dispose of contents/container in accordance with local/regional/national/international regulations.}]
[{code=MIXTURE LIST, sequence=1, description=Contains : nitrocellulose}]
[{code=GHS02, sequence=1, description=FLAME}]
Product Identifier
  • X100 MEMBRANFILTER AE98 47MM,
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H226- Flammable liquid and vapour.
Precautionary Statement
  • P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241- Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260- Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280- Wear protective gloves/protective clothing/eye protection/face protection.
  • P501b- Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental Information
  • MIXTURE LIST- Contains : nitrocellulose