Pyrex Pyrex™ Borosilicate Glass Cylinder with Polyethylene Foot
Manufactured from Pyrex™ borosilicate glass and polyethylene for strength, stability and superior chemical resistance. Pyrex™ Borosilicate Glass Cylinder with Polyethylene Foot is an excellent device for measuring and mixing liquids in a laboratory.
Overview
22.83 GBP valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Code: 01
Additional Details:
Additional Details: Weight: 0.31950kg
Product Code. 11363564
Quantity Price
1 £ 54.55 / Pack of 2
In stock, ready to ship: 3
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
  • Accessories and Spare Parts
Description and Specification
Specification
Base Type Hexagonal
Scale Color White
Shape Round
Type Graduated Cylinder
Tolerance ±1 mL
Material Borosilicate Glass
Certifications/Compliance BS 604, DIN 12680, ISO 4788
Class B
Capacity (Metric) 100 mL
Height (Metric) 270 mm
Includes Polyethylene Foot
Scale White enamel
Graduations 1 mL
  • Borosilicate glass cylinder: white enamel graduations for optimal visibility
  • Polyethylene foot: absorbs impact and reduces breakage
  • Class B
  • Complies with: DIN 12680, BS 604, ISO 4788
  • Replacement feet: available separately