Fisherbrand™ Bead Mill 24 Homogenizer
Used for variety of applications that require grinding, lysing, and homogenization of biological samples prior to molecular extraction. Used with Fisherbrand™ Bead Kits.
Overview
7604.36 GBP valid until 2025-12-31
Use promo code "25045" to get your promotional price.
Additional Details:
Additional Details: Weight: 50.00000kg
Product Code. 15515799
Quantity Price
1 £ 8565.0 / Each
Estimated Shipment
02-06-2025
Add to basket
  • Description and Specification
  • Documentation
  • Accessories and Spare Parts
Description and Specification
Specification
Product Type Homogenizer
Electrical Requirements 110-230 VAC/50-60 Hz
Dimensions (D x W x H) 17 x 15 x 13 in.
For Use With (Application) DNA/RNA extraction, tissue homogenization, protein purification
Weight (English) 65 lb.
Weight (Metric) 29.5 kg
  • Simultaneously homogenize up to 24 x 2 mL or 0.5 mL tubes
  • More samples can be processed in smaller amount of time
  • Convenient front-loading tube holder position for ease of use and optimal bead/sample interactions
  • Includes 24 place tube carriage and Bead Mill Finger Plate that holds the 2mL tubes firmly in place during processing
  • Processing range is ideal for soft tissue samples
  • No cool-down required between runs (non-stop processing)
  • Disposable tubes ensure no threat of cross-contamination
  • No sample degradation due to quick processing
  • Smaller footprint to maximize bench space
  • Run Time: 1 to 99 sec.
  • Performance Range: 0.8 m/s to 6 m/s in increments of 1.5 m/s
  • Number of Cycles: 1-9
Compliance: CE compliant
Documentation Accessories and Spare Parts