Quickfit Quickfit™ Three-Vertical-Neck Round-Bottom Flask
Includes three necks for easy addition of measurement tools. The Quickfit™ Three-Vertical-Neck Round-Bottome Flask is made from borosilicate glass.
Overview
430.13 GBP valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Code: NEW
Additional Details:
Additional Details: Weight: 0.35000kg
Product Code. 15527860
Quantity Price
1 £ 648.0 / Pack of 5
Estimated Shipment
22-08-2025
Add to basket
Description and Specification
Specification
Capacity (Metric) 1000 mL
Material Borosilicate Glass
Height (Metric) 180 mm
No. per Case 3
Socket Joint 24/29 (Centre), 19/26 (side)
Bottom Shape Round
  • Fitted with three Quickfit™ precision-ground sockets - One vertical centre neck - Two angled side necks
  • Quickfit™ ground glass sockets are manufactured to an accuracy which surpasses the requirements of ISO 383
Documentation