Code:
NEW
Honeywell Fluka Buffer Solution pH 7.0 (20°C), Potassium Dihydrogen Phosphate / Disodium Hydrogen Phosphate, Traceable to SRM from NIST, with Fungicide, Honeywell Fluka™
Potassium dihydrogen phosphate / disodium hydrogen phosphate, traceable to SRM from NIST, with fungicide.
Additional Details:
CAS Number:
7778-77-0
Weight:
2.98500kg
Quantity | 6 x 500 mL |
---|
Chemical Identifiers
CAS | |
---|---|
Molecular Formula | |
Molecular Weight (g/mol) | |
InChI Key | |
Synonym | Potassium sodium phosphate buffer solution |
PubChem CID | |
ChEBI | |
SMILES |
Specification
Chemical Name or Material | Buffer solution pH 7.0 (20°C) |
---|---|
Name Note | Potassium dihydrogen phosphate / disodium hydrogen phosphate, traceable to SRM from NIST, with fungicide |
Boiling Point | 100°C |
CAS Min % | ≥90.0000% |
CAS Max % | ≤100.0000% |
CAS | 7558-79-4 |
CAS Max % | 0.7300% |
Density | 1 g/cm3 |
CAS | 7778-77-0 |
CAS Max % | 0.3500% |
Quantity | 6 x 500 mL |
Solution Type | Buffer |
MDL Number | MFCD00146206 |
Packaging | Poly Bottle |
Synonym | Potassium sodium phosphate buffer solution |