OHAUS™ Incubating Shaker
Save Incubator Space with Benchtop Incubating and Incubating Refrigerating Shakers
4919.56 GBP valid until 2025-12-31
Use promo code "24360" to get your promotional price.
Use promo code "24360" to get your promotional price.
Additional Details:
Weight:
35.90000kg
Specification
Load Bearing Capacity (Metric) | 16 kg at 75 rpm, 2.3 kg at 500 rpm |
---|---|
Speed Range | 600 to 2500 rpm |
Voltage | 230 V |
Temperature Range (Metric) | 4°C to 40°C |
Wattage | 450 W |
Load Bearing Capacity (English) | 35 lb. at 75 rpm, 5 lb. at 500 rpm |
Certifications/Compliance | cULus |
Frequency | 50/60 Hz |
Material | Aluminum, Cycoloy |
Dimensions (L x W x H) | 648 x 356 x 406 mm |
Height (English) | 16 in. |
Length (English) | 25.5 in. |
Width (English) | 14 in. |
Shipping Weight (Metric) | 37.7 kg |
Weight (English) | 79.11 lb. |
Weight (Metric) | 35.9 kg |
Model | ISHD16HDG |
Display | Independent LED displays for temperature, speed and time |
For Use With (Application) | Cell Cultures, Solubility Studies, Bacterial Suspensions, Extraction Procedures |
Humidity Range | 20 to 80%, Non-condensing |
Timer Range | 1 sec. to 160 hr. |